N,N'-Diallyl-L-tartardiamide ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12195.18
A12195.30
A12195.18EA
2360
CZK
A12195.18
A12195.30
N,N'-Diallyl-L-tartardiamide ≥99%
N,N'-Diallyl-L-tartardiamide
Purity: 99%
CAS No.: 58477-85-3
Molecular Formula: C10H16N2O4
Formula Weight: 228.25
Melting Point: 184-186°
Beilstein Reference: 1712934
Vzorec:
C₁₀H₁₆N₂O₄ Bod tání: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008640 CAS číslo: 58477-85-3 EINECS: 261-277-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...