DL-Methionine sulphoxide
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
H-Met(O)-OH, (2S)-2-Amino-4-(methylsulphinyl)butanoic acid
J62873.03
J62873.06
J62873.14
J62873.03EA
1480
CZK
J62873.03
J62873.06
J62873.14
DL-Methionine sulphoxide
L-Methionine sulphoxide
CAS No.: 3226-65-1
Molecular Formula: C5H11NO3S
Formula Weight: 165.21
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4129939
MDL No.: MFCD00063093
Vzorec:
C₅H₁₁NO₃S MW: 165.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00063093 CAS číslo: 3226-65-1 EINECS: 221-758-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...