Tisk…

Thiodiglycolic anhydride ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1,4-Oxathiane-2,6-dione

A13390.06 A13390.22
A13390.06EA 1920 CZK
A13390.06 A13390.22
Thiodiglycolic anhydride ≥98%
Thiodiglycolic anhydride

Purity: 98% CAS No.: 3261-87-8 Molecular Formula: C4H4O3S Formula Weight: 132.13 Melting Point: 95-100° Beilstein Reference: 112528

Vzorec: C₄H₄O₃S
MW: 132,13 g/mol
Bod tání: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00051689
CAS číslo: 3261-87-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR