Tisk…

6-Indoleboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

666459-1G 666459-5G
SIAL666459-1GEA 2770 CZK
SIAL666459-1G SIAL666459-5G
6-Indoleboronic acid, Sigma-Aldrich®
6-Indoleboronic acid
Vzorec: C₈H₈BNO₂
MDL Number: MFCD03095176
CAS číslo: 147621-18-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR