Tisk…

6-Indoleboronic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H53313.MD H53313.03
H53313.MDEA 3040 CZK
H53313.MD H53313.03
6-Indoleboronic acid ≥98%
6-Indoleboronic acid

Purity: 98% CAS No.: 147621-18-9 Molecular Formula: C8H8BNO2 Formula Weight: 160.97 Storage Temperature: -30°C to -10°C Melting Point: 177-181° MDL No.: MFCD03095176

Vzorec: C₈H₈BNO₂
Bod tání: 177…181 °C
Storage Temperature: Freezer
MDL Number: MFCD03095176
CAS číslo: 147621-18-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR