5-Indoleboronic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid
359461000
359465000
359460010
ACRO359461000EA
920
CZK
ACRO359461000
ACRO359465000
ACRO359460010
5-Indoleboronic acid 97%
5-Indoleboronic acid
Vzorec:
C₈H₈BNO₂ MW: 160,97 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01319013 CAS číslo: 144104-59-6 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | Light yellow to beige Powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...