Tisk…

1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

654280-1G 654280-5G
SIAL654280-1GEA 4650 CZK
SIAL654280-1G SIAL654280-5G
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester, Sigma-Aldrich®
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester
Vzorec: C₂₀H₂₂BNO₄S
MDL Number: MFCD08276778
CAS číslo: 870717-93-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR