Tisk…

Dibenzyl phosphate ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Dibenzyl hydrogen phosphate

L08813.03 L08813.06 L08813.14
L08813.03EA 858 CZK
L08813.03 L08813.06 L08813.14
Dibenzyl phosphate ≥98%
Dibenzyl phosphate

Purity: 98% CAS No.: 1623-08-1 Molecular Formula: C14H15O4P Formula Weight: 278.24 Storage Temperature: 2°C to 8°C Melting Point: 78-81° Beilstein Reference: 2055755 MDL No.: MFCD00004775

Vzorec: C₁₄H₁₅O₄P
MW: 278.24 g/mol
Bod tání: 78…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004775
CAS číslo: 1623-08-1
EINECS: 216-602-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR