Tisk…

Dibenzyl phosphate 98%

Dodavatel: Thermo Scientific

Synonyms: Dibenzyl hydrogen phosphate

112650250 112650050 112650010
ACRO112650250EA 9860 CZK
ACRO112650250 ACRO112650050 ACRO112650010
Dibenzyl phosphate 98%
Dibenzyl phosphate
Vzorec: C₁₄H₁₅O₄P
MW: 278,24 g/mol
Bod tání: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004775
CAS číslo: 1623-08-1
EINECS: 216-602-3

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
Titration with NaOH ≥97.5 %
Sulfate (SO4) Passes test (limit about 0.002%)
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤20 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR