Tisk…

Furfuryl methyl sulphide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-((Methylthio)methyl)furan

M-Clarity™ quality level = MQ300

W316008-100G-K W316008-1KG-K
SIALW316008-100G-KEA 5900 CZK
SIALW316008-100G-K SIALW316008-1KG-K
Furfuryl methyl sulphide, Sigma-Aldrich®
Furfuryl methyl sulphide
Vzorec: C₆H₈OS
MW: 128,19 g/mol
MDL Number: MFCD00009604
CAS číslo: 1438-91-1
EINECS: 215-874-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR