Difurfuryl sulphide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W323802-100G
W323802-25G
W323802-SAMPLE
SIALW323802-100GEA
8150
CZK
SIALW323802-100G
SIALW323802-25G
SIALW323802-SAMPLE
Difurfuryl sulphide, Sigma-Aldrich®
Difurfuryl sulphide
Vzorec:
C₁₀H₁₀O₂S |
MDL Number:
MFCD00010081 CAS číslo: 13678-67-6 EINECS: 237-172-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...