Tisk…

Ebselen, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

M-Clarity™ quality level = MQ200

E3520-100MG E3520-25MG
SIALE3520-100MGEA 22720 CZK
SIALE3520-100MG SIALE3520-25MG
Ebselen, Sigma-Aldrich®
Ebselen
Vzorec: C₁₃H₉NOSe
MW: 274.20 g/mol
MDL Number: MFCD00210937
CAS číslo: 60940-34-3
UN: 3283
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR