Tisk…

Zirconium diboride ≥99.5% (metals basis excluding Hf)

Dodavatel: Thermo Scientific
Varování

Synonyms: Zirconium boride

12140.18 12140.30
12140.18EA 4840 CZK
12140.18 12140.30
Zirconium diboride ≥99.5% (metals basis excluding Hf)
Zirconium diboride

Purity: 99.5% CAS No.: 12045-64-6 Molecular Formula: ZrB2 Formula Weight: 112.84 Physical Form: -325 Mesh Powder Melting Point: 3000° UN No.: 3178 MDL No.: MFCD00064648

Vzorec: B₂Zr
MW: 112,84 g/mol
Bod tání: 3000 °C
Storage Temperature: Ambient
MDL Number: MFCD00064648
CAS číslo: 12045-64-6
EINECS: 234-963-5
UN: 3178
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR