4-Methoxy-2-(trifluoromethyl)phenylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Methoxy-2-(trifluoromethyl)benzeneboronic acid, (4-Methoxy-2-(trifluoromethyl)phenyl)boronic acid
M-Clarity™ quality level = MQ100
666912-1G
666912-5G
SIAL666912-1GEA
0
CZK
SIAL666912-1G
SIAL666912-5G
4-Methoxy-2-(trifluoromethyl)phenylboronic acid, Sigma-Aldrich®
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
Vzorec:
C₈H₈BF₃O₃ MW: 219,95 g/mol |
MDL Number:
MFCD03095346 CAS číslo: 313546-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...