Tisk…

4-Chloro-3-methylphenylboronic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Chloro-3-methylbenzeneboronic acid

B23179.03 B23179.06
B23179.03EA 2080 CZK
B23179.03 B23179.06
4-Chloro-3-methylphenylboronic acid ≥98%
4-Chloro-3-methylphenylboronic acid

Purity: 98% CAS No.: 161950-10-3 Molecular Formula: C7H8BClO2 Formula Weight: 170.40 Melting Point: 238-242° MDL No.: MFCD02683104

Vzorec: C₇H₈BClO₂
MW: 170.40 g/mol
Bod tání: 238…242 °C
Storage Temperature: Ambient
MDL Number: MFCD02683104
CAS číslo: 161950-10-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR