Tisk…

3-Chloro-4-methylphenylboronic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid

H28288.06 H28288.03
H28288.06EA 2460 CZK
H28288.06 H28288.03
3-Chloro-4-methylphenylboronic acid ≥97%
3-Chloro-4-methylphenylboronic acid

Purity: 97% CAS No.: 175883-63-3 Molecular Formula: C7H8BClO2 Formula Weight: 170.40 Melting Point: 224-230° MDL No.: MFCD04039010

Vzorec: C₇H₈BClO₂
MW: 170,40 g/mol
Bod tání: 224…230 °C
Storage Temperature: Ambient
MDL Number: MFCD04039010
CAS číslo: 175883-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR