Tisk…
Flame-retardant rain trousers, Ekofisk 5806
Dodavatel: SIOEN
5806/B75/1XL 5806/B75/2XL 5806/B75/3XL 5806/B75/L 5806/B75/M 5806/B75/S
113-7116EA 5640 CZK
113-7116 113-7117 113-7118 113-7119 113-7120 113-7121
Flame-retardant rain trousers, Ekofisk 5806
Pracovní oděvy
These lightweight trousers made of Siopor® FR AST fabric (100% polyester fabric with 100% FR PU coating + AST) are waterproof and windproof and have anti-static features. The fixed cotton lining is flame-retardant.
  • High tear resistance, taped seams
  • Front fly with press studs, two side slits
  • Ankles adjustable by touch and close fastening, elasticated waist with drawstring
  • Hidden zip closure in trouser legs
  • Ideal for combination with Hasnon jacket
The trousers provide good protection against rough weather conditions.
Weight: 250 g/m²
Certifikace: More Product Information
Objednat
Produktová specifikace
Model Ekofisk 5806
Environmentally Preferable
Aplikace oblečení Fire protection
Risk area Rain (waterproof),Flame-retardant,Anti-static
Rod Pro muže
Sleeve or leg length Long
Typ uzávěru Zapínání na druky
Tkanina 100% polyester fabric with 100% FR PU coating + AST
Weight 250 g/m²