Tisk…
SPE columns, PCB analysis, CHROMABOND® NAN
Dodavatel: MACHEREY-NAGEL
730149 730109 730149G 730149.250
MANA730149EA 11880 CZK
MANA730149 MANA730109 MANA730149G MANA730149.250
SPE columns, PCB analysis, CHROMABOND® NAN
Kolony chromatografické
CHROMABOND® NAN is a special combination phase for SPE analysis of PCBs from sludge.
  • Sodium sulfate for removal of trace water (N)
  • SiOH/AgNO₃ phase for removal of sulfur, sulfur-containing and polar compounds (A)
Application: PCB extraction from sludge (hexane extract).