Acetaldehyd-D4 (98% D), Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
176567-1G
176567-5G
SIAL176567-1GEA
5870
CZK
SIAL176567-1G
SIAL176567-5G
Acetaldehyd-D4 (98% D), Sigma-Aldrich®
Acetaldehyd-D4
Vzorec:
C₂D₄O Storage Temperature: Refrigerator |
MDL Number:
MFCD00044230 CAS číslo: 1632-89-9 EINECS: 216-641-6 UN: 1089 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...