4'-Fluorobiphenyl-4-carboxylic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid, 4-(4-Fluorophenyl)benzoic acid
H33208.03
H33208.06
H33208.03EA
3010
CZK
H33208.03
H33208.06
4'-Fluorobiphenyl-4-carboxylic acid 96%
4'-Fluorobiphenyl-4-carboxylic acid
Vzorec:
C₁₃H₉FO₂ MW: 216.21 Bod tání: 243…244 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631909 CAS číslo: 5731-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...