Tisk…

2-Amino-4-phenylthiazole

Dodavatel: Cayman Chemical

Synonyms: 4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine

13388-500 13388-1 13388-5 13388-10
CAYM13388-500EA 920 CZK
CAYM13388-500 CAYM13388-1 CAYM13388-5 CAYM13388-10
2-Amino-4-phenylthiazole
2-Amino-4-phenylthiazole
Vzorec: C₉H₈N₂S
MDL Number: MFCD00039680
CAS číslo: 2010-06-2
EINECS: 217-926-8
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR