1-Bromo-3-methyl-2-butene stabilizovaný, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,3-Dimethylallyl bromide, 4-Bromo-2-methyl-2-butene
Stabilization:
Stabilised with silver
M-Clarity™ quality level = MQ200
274372-10G
274372-1G
SIAL274372-10GEA
2580
CZK
SIAL274372-10G
SIAL274372-1G
1-Bromo-3-methyl-2-butene stabilizovaný, Sigma-Aldrich®
1-Bromo-3-methyl-2-butene
Vzorec:
C₅H₉Br MW: 149,03 g/mol Bod varu: 129…133 °C (1013 hPa) Hustota: 1,293 g/cm³ (15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000242 CAS číslo: 870-63-3 EINECS: 212-799-5 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...