Tisk…

4-Bromo-3-fluorobenzaldehyde ≥97%

Dodavatel: Thermo Scientific
Varování

H57833.03 H57833.06
H57833.03EA 2730 CZK
H57833.03 H57833.06
4-Bromo-3-fluorobenzaldehyde ≥97%
4-Bromo-3-fluorobenzaldehyde

Purity: 97% CAS No.: 133059-43-5 Molecular Formula: C7H4BrFO Formula Weight: 203.01 Melting Point: 54-60° Beilstein Reference: 9321564

Vzorec: C₇H₄BrFO
MW: 203,01 g/mol
Bod tání: 54…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03095000
CAS číslo: 133059-43-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR