Dichloro(methyl)silane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methyldichlorosilane
M-Clarity™ quality level = MQ200
440248-100ML
440248-1L
SIAL440248-100MLEA
1060
CZK
SIAL440248-100ML
SIAL440248-1L
Dichloro(methyl)silane, Sigma-Aldrich®
Dichloro(methyl)silane
Vzorec:
CH₄Cl₂Si MW: 115,03 g/mol Bod varu: 41 °C (1013 hPa) Bod tání: –92,8 °C Hustota: 0,93 g/cm³ (0 °C) |
MDL Number:
MFCD00000494 CAS číslo: 75-54-7 EINECS: 200-877-1 UN: 1242 ADR: 4.3,I REACH: 01-2119436669-21 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...