Tisk…

Cyclopropanesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cyclopropanesulphonylchloride

M-Clarity™ quality level = MQ100

640573-100MG 640573-500MG
SIAL640573-100MGEA 2320 CZK
SIAL640573-100MG SIAL640573-500MG
Cyclopropanesulphonyl chloride, Sigma-Aldrich®
Cyclopropanesulphonyl chloride
Vzorec: C₃H₅ClO₂S
MDL Number: MFCD01631933
CAS číslo: 139631-62-2
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR