Indium(III) chloride hydrate ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
11859.09
11859.18
11859.30
11859.09EA
2790
CZK
11859.09
11859.18
11859.30
Indium(III) chloride hydrate ≥99.99% (metals basis)
Indium(III) chloride hydrate
Purity: 99.99%
CAS No.: 143983-91-9
Molecular Formula: InCl3.xH2O
Formula Weight: 221.18(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Solubility: Soluble in water
Merck Reference: 14,4958
UN No.: 3260
MDL No.: MFCD00011058
Vzorec:
Cl₃In·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149744 CAS číslo: 143983-91-9 EINECS: 233-043-0 UN: 3260 ADR: 8,II Merck Index: 14,4958 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...