Tisk…

Methyl-5-bromosalicylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 5-bromo-2-hydroxybenzoate

M-Clarity™ quality level = MQ200

124621-25G 124621-5G
SIAL124621-25GEA 7410 CZK
SIAL124621-25G SIAL124621-5G
Methyl-5-bromosalicylate, Sigma-Aldrich®
Methyl-5-bromosalicylate
Vzorec: C₈H₇BrO₃
MW: 231,05 g/mol
MDL Number: MFCD00016460
CAS číslo: 4068-76-2
EINECS: 223-776-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR