(±)-Aspartic acid ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
2-Aminobutanedioic acid, DL-Aspartic acid
A13646.0E
A13646.22
A13646.36
A13646.0EEA
8550
CZK
A13646.0E
A13646.22
A13646.36
(±)-Aspartic acid ≥98%
(±)-Aspartic acid
Purity: 98+%
CAS No.: 617-45-8
Molecular Formula: C4H7NO4
Formula Weight: 133.10
Melting Point:
Vzorec:
C₄H₇NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00063083 CAS číslo: 617-45-8 EINECS: 210-513-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...