DL-Glutamic acid monohydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
DL-Glutamic Acid Monohydrate
M-Clarity™ quality level = MQ200
G1126-100G
G1126-1KG
G1126-25G
SIALG1126-100GEA
8070
CZK
SIALG1126-100G
SIALG1126-1KG
SIALG1126-25G
DL-Glutamic acid monohydrate, Sigma-Aldrich®
DL-Glutamic acid monohydrate
Vzorec:
C₅H₉NO₄·1H₂O MW: 165,15 g/mol (147,13 anhy) |
MDL Number:
MFCD00150703 CAS číslo: 19285-83-7 EINECS: 210-522-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...