DL-Glutamic acid monohydrate ≥99%
Dodavatel: Thermo Fisher Scientific
A17719.14
A17719.22
A17719.36
A17719.14EA
1010
CZK
A17719.14
A17719.22
A17719.36
DL-Glutamic acid monohydrate ≥99%
DL-Glutamic acid monohydrate
Purity: 99%
CAS No.: 19285-83-7
Molecular Formula: C5H9NO4.H2O
Formula Weight: 165.15 (147.13anhy)
Melting Point: ca 200° dec.
Beilstein Reference: 4155097
Merck Reference: 14,4469
MDL No.: MFCD00150703
Vzorec:
C₅H₉NO₄·1H₂O Hustota: 1,460 Storage Temperature: Ambient |
MDL Number:
MFCD00150703 CAS číslo: 19285-83-7 EINECS: 210-522-2 Merck Index: 14,4469 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...