DL-Glutamic acid monohydrate 98+%
Dodavatel: Thermo Fisher Scientific
Synonyms:
DL-Glutamic Acid Monohydrate
119930010
119931000
119930250
ACRO119930010EA
13020
CZK
ACRO119930010
ACRO119931000
ACRO119930250
DL-Glutamic acid monohydrate 98+%
DL-Glutamic acid monohydrate
Vzorec:
C₅H₉NO₄·1H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150703 CAS číslo: 19285-83-7 EINECS: 210-522-2 |
Výsledky specifikačního testu
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤15.5 % (K.F.) |
Titration with NaOH | ≥98.0 % |
Specific optical rotation | +31° to +35° (20°C, 589 nm) (c=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...