Tisk…

Homophthalic anhydride ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A15538.09 A15538.18
A15538.09EA 1860 CZK
A15538.09 A15538.18
Homophthalic anhydride ≥97%
Homophthalic anhydride

Purity: 97+% CAS No.: 703-59-3 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 140-144° Beilstein Reference: 5244

Vzorec: C₉H₆O₃
MW: 162.14 g/mol
Bod tání: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00006894
CAS číslo: 703-59-3
EINECS: 211-873-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR