Tisk…
1-Bromo-2-cyclohexylbenzene ≥97%
Dodavatel: Thermo Fisher Scientific
L18059.14 L18059.06 L18059.03
L18059.14EA 18810 CZK
L18059.14 L18059.06 L18059.03
1-Bromo-2-cyclohexylbenzene ≥97%
1-Bromo-2-cyclohexylbenzene
Purity: 97% CAS No.: 59734-92-8 Molecular Formula: C12H15Br Formula Weight: 239.16 Refractive Index: 1.5605 Boiling Point: 79-83°/0.01mm MDL No.: MFCD02093987
Vzorec: C₁₂H₁₅Br
Bod varu: 79…83 °C (0,01 mmHg)
Hustota: 1,295
Storage Temperature: Ambient
MDL Number: MFCD02093987
CAS číslo: 59734-92-8