Benzyl methyl ether ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(Methoxymethyl)benzene
B23490.09
B23490.18
B23490.09EA
1410
CZK
B23490.09
B23490.18
Benzyl methyl ether ≥97%
Benzyl methyl ether
Purity: 99%
CAS No.: 538-86-3
Molecular Formula: C6H5CH2OCH3
Formula Weight: 122.17
Flash Point: 56°(133°F)
Refractive Index: 1.5015
Melting Point: -53°
Boiling Point: 173-175°
Merck Reference: 14,1140
UN No.: 3271
MDL No.: MFCD00025901
Vzorec:
C₈H₁₀O MW: 122,17 g/mol Bod varu: 173…175 °C Bod tání: –53 °C Hustota: 0,940 Bod vzplanutí: 56 °C (133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025901 CAS číslo: 538-86-3 EINECS: 208-705-7 UN: 3271 ADR: 3,III Merck Index: 14,1140 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...