Tisk…

1-Bromo-3-hexylbenzene ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H26063.14 H26063.06
H26063.14EA 8070 CZK
H26063.14 H26063.06
1-Bromo-3-hexylbenzene ≥97%
1-Bromo-3-hexylbenzene

Purity: 97+% CAS No.: 38409-59-5 Molecular Formula: C12H17Br Formula Weight: 241.17 Flash Point: 120°(248°F) Refractive Index: 1.5220 Boiling Point: 90-98°/5mm MDL No.: MFCD09909567

Vzorec: C₁₂H₁₇Br
MW: 241.17 g/mol
Bod varu: 90…98 °C (5 mmHg)
Hustota: 1.179
Bod vzplanutí: >120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD09909567
CAS číslo: 38409-59-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR