Tisk…

1-Dodecylmagnesium bromide 1 mol/l in diethyl ether, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

339628-100ML 339628-800ML
SIAL339628-100MLEA 2780 CZK
SIAL339628-100ML SIAL339628-800ML
1-Dodecylmagnesium bromide 1 mol/l in diethyl ether, Sigma-Aldrich®
1-Dodecylmagnesium bromide
Vzorec: C₁₂H₂₅BrMg
MDL Number: MFCD00075165
CAS číslo: 15890-72-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR