Tisk…

Methyl (4-methoxyphenyl)acetate ≥97%

Dodavatel: Thermo Fisher Scientific

A17940.14 A17940.22 A17940.36
A17940.14EA 1480 CZK
A17940.14 A17940.22 A17940.36
Methyl (4-methoxyphenyl)acetate ≥97%
Methyl (4-methoxyphenyl)acetate

Purity: 97+% CAS No.: 23786-14-3 Molecular Formula: C10H12O3 Formula Weight: 180.23 Flash Point: 100°(212°F) Refractive Index: 1.5160 Boiling Point: 157-159°/19mm Beilstein Reference: 2209665 MDL No.: MFCD00008454

Vzorec: C₁₀H₁₂O₃
Bod varu: 157…159 °C (19 mmHg)
Hustota: 1,135
Bod vzplanutí: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008454
CAS číslo: 23786-14-3
EINECS: 245-886-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR