Tisk…

3-Methoxypropionic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Methoxypropanoic acid

M-Clarity™ quality level = MQ200

374547-10ML 374547-50ML
SIAL374547-10MLEA 6790 CZK
SIAL374547-10ML SIAL374547-50ML
3-Methoxypropionic acid, Sigma-Aldrich®
3-Methoxypropionic acid
Vzorec: C₄H₈O₃
MW: 104,1100 g/mol
MDL Number: MFCD00042654
CAS číslo: 2544-06-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR