3-Bromo-4-heptanone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H32094.03
H32094.09
H32094.03EA
1900
CZK
H32094.03
H32094.09
3-Bromo-4-heptanone ≥98%
3-Bromo-4-heptanone
Purity: 98%
CAS No.: 42330-10-9
Molecular Formula: C7H13BrO
Formula Weight: 193.08
Flash Point: 75°(167°F)
Refractive Index: 1.4580
Boiling Point: 39°/1mm
UN No.: 3265
MDL No.: MFCD09757526
Vzorec:
C₇H₁₃BrO Bod varu: 39 °C (1 mmHg) Hustota: 1,248 Bod vzplanutí: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD09757526 CAS číslo: 42330-10-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...