křemík, powder, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
633097-10G
633097-25G
SIAL633097-10GEA
29950
CZK
SIAL633097-10G
SIAL633097-25G
křemík, powder, Sigma-Aldrich®
křemík
Vzorec:
Si MW: 28.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS číslo: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1 III Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...