Tisk…

2,5-Dichloro-3-thiophenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

544280-1G 544280-5G
SIAL544280-1GEA 0 CZK
SIAL544280-1G SIAL544280-5G
2,5-Dichloro-3-thiophenesulphonyl chloride, Sigma-Aldrich®
2,5-Dichloro-3-thiophenesulphonyl chloride
Vzorec: C₄HCl₃O₂S₂
MW: 251,54 g/mol
MDL Number: MFCD00051665
CAS číslo: 56946-83-9
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR