Tisk…

(±)-3-Amino-3-phenylpropionic acid 99%

Dodavatel: Thermo Scientific

Synonyms: H-DL-β-Phe-OH

L19801.03 L19801.06
L19801.03EA 814 CZK
L19801.03 L19801.06
(±)-3-Amino-3-phenylpropionic acid 99%
(±)-3-Amino-3-phenylpropionic acid

Purity: 99% CAS No.: 614-19-7 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Storage Temperature: 2°C to 8°C Melting Point: ca 220° dec. Beilstein Reference: 2803286 MDL No.: MFCD00008064

Vzorec: C₉H₁₁NO₂
MW: 165,19 g/mol
Bod tání: ∼ 220 °C (rozkl.)
MDL Number: MFCD00008064
CAS číslo: 614-19-7
EINECS: 210-371-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR