Ethyl (2-bromophenyl)acetate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
H50630.03
H50630.MD
H50630.03EA
3920
CZK
H50630.03
H50630.MD
Ethyl (2-bromophenyl)acetate ≥98%
Ethyl (2-bromophenyl)acetate
Purity: 98%
CAS No.: 2178-24-7
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Melting Point: 35-38°
MDL No.: MFCD00051921
Vzorec:
C₁₀H₁₁BrO₂ MW: 243,10 g/mol Bod tání: 35…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051921 CAS číslo: 2178-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...