2,2,2-Trifluoroethanol-[D₂] (99% D), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,2,2-Trifluoroethanol-[²H₂]
M-Clarity™ quality level = MQ200
426237-25ML
426237-5ML
SIAL426237-25MLEA
15480
CZK
SIAL426237-25ML
SIAL426237-5ML
2,2,2-Trifluoroethanol-[D₂] (99% D), Sigma-Aldrich®
2,2,2-Trifluoroethanol-[D₂]
Vzorec:
F₃CCD₂OD Storage Temperature: Ambient |
CAS číslo:
77568-66-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...