3-Bromo-4-nitropyridine-N-oxide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Bromo-4-nitropyridine 1-oxide
L19524.14
L19524.06
L19524.14EA
12050
CZK
L19524.14
L19524.06
3-Bromo-4-nitropyridine-N-oxide ≥98%
3-Bromo-4-nitropyridine-N-oxide
Purity: 98+%
CAS No.: 1678-49-5
Molecular Formula: C5H3BrN2O3
Formula Weight: 219.00
Melting Point: 152-154°
Beilstein Reference: 150822
Vzorec:
C₅H₃BrN₂O₃ MW: 219.00 g/mol Bod tání: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085925 CAS číslo: 1678-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...