Trimethylphosphine oxide
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Trimethylphosphineoxide
30394.03
30394.06
30394.03EA
5190
CZK
30394.03
30394.06
Trimethylphosphine oxide
Trimethylphosphine oxide
CAS No.: 676-96-0
Molecular Formula: (CH3)3P(O)
Formula Weight: 92.08
Physical Form: Crystalline
Odour: Odourless
Melting Point: 140-141°
MDL No.: MFCD00013910
Vzorec:
C₃H₉OP Bod tání: 140…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013910 CAS číslo: 676-96-0 EINECS: 211-633-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...