4-Amino-α,α,α-trifluoro-m-toluic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Amino-3-(trifluoromethyl)benzoic acid
H32487.03
H32487.09
H32487.03EA
714
CZK
H32487.03
H32487.09
4-Amino-α,α,α-trifluoro-m-toluic acid 98%
4-Amino-α,α,α-trifluoro-m-toluic acid
Purity: 98%
CAS No.: 400-76-0
Molecular Formula: C8H6F3NO2
Formula Weight: 205.14
Melting Point: 212-215°
Beilstein Reference: 3291493
Vzorec:
C₈H₆F₃NO₂ MW: 205,14 g/mol Bod tání: 212…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD03407960 CAS číslo: 400-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...