Tisk…

4-Amino-α,α,α-trifluoro-o-toluic acid ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Amino-2-(trifluoromethyl)benzoic acid

H32626.03 H32626.06
H32626.03EA 622 CZK
H32626.03 H32626.06
4-Amino-α,α,α-trifluoro-o-toluic acid ≥97%
4-Amino-α,α,α-trifluoro-o-toluic acid

Purity: 97+% CAS No.: 393-06-6 Molecular Formula: C8H6F3NO2 Formula Weight: 205.14 Melting Point: 183-186° MDL No.: MFCD03407959

Vzorec: C₈H₆F₃NO₂
MW: 205,14 g/mol
Bod tání: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD03407959
CAS číslo: 393-06-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR