Tisk…

2-Ethynylanisole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2'-Methoxyphenyl acetylene, 1-Ethynyl-2-methoxybenzene

467227-1G 467227-5G
SIAL467227-1GEA 2470 CZK
SIAL467227-1G SIAL467227-5G
2-Ethynylanisole, Sigma-Aldrich®
2-Ethynylanisole
Vzorec: C₉H₈O
MW: 132,1600 g/mol
MDL Number: MFCD03839989
CAS číslo: 767-91-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR