Tisk…

2-Amino-5-(trifluoromethoxy)benzoic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

H32916.03 H32916.06
H32916.03EA 3120 CZK
H32916.03 H32916.06
2-Amino-5-(trifluoromethoxy)benzoic acid 98%
2-Amino-5-(trifluoromethoxy)benzoic acid

Purity: 98% CAS No.: 83265-56-9 Molecular Formula: C8H6F3NO3 Formula Weight: 221.14 MDL No.: MFCD00176651

Vzorec: C₈H₆F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00176651
CAS číslo: 83265-56-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR